missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ ZWITTERGENT™ 3-12 Detergent, Calbiochem™,
$227.00 - $6,060.00
Chemical Identifiers
| CAS | 14933-08-5 |
|---|---|
| Molecular Formula | C17H37NO3S |
| Molecular Weight (g/mol) | 335.547 |
| InChI Key | IZWSFJTYBVKZNK-UHFFFAOYSA-N |
| Synonym | n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
| PubChem CID | 84703 |
| ChEBI | CHEBI:75303 |
| IUPAC Name | 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate |
| SMILES | CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6930155GM
|
MilliporeSigma™
6930155GM |
5 g | Plastic Bags |
N/A
|
|
||||
|
69301525GM
|
MilliporeSigma™
69301525GM |
25 g | Glass Bottle |
N/A
|
|
||||
|
6930151KG
|
MilliporeSigma™
6930151KG |
1 kg | Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
| 14933-08-5 | |
| 335.547 | |
| n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| CHEBI:75303 | |
| CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| C17H37NO3S | |
| IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
| 84703 | |
| 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate |
Specifications
| White | |
| 5 g | |
| C17H37NO3S | |
| IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
| 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate | |
| 84703 | |
| ≥99% (TLC) |
| 14933-08-5 | |
| Plastic Bags | |
| n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 335.547 | |
| CHEBI:75303 |