missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Acrylic Burette with PTFE Key Stopcock, Class B

Transparent acrylic burette features easy to read graduations and leakproof PTFE plunger stopcock.

Supplier:  Eisco™ CH0233A

 View more versions of this product

Catalog No. S27028


Explore available promotions

Description

Description

productFeatures
  • Made of high quality acrylic, highly resistant to impact and shattering
  • Acrylic body is resistant to dilute minerals, acids and bases except hydrofluoric acid and ammonium hydroxide
  • Includes interchangeable, nonstick, chemical resistant PTFE key stopcock
  • Calibrated to deliver
  • Clean with non-abrasive cleaners, including strong detergents
  • Do not autoclave
  • Replacement stopcocks available; S24498
Specifications

Specifications

0.1 mL
Key Stopcock
PTFE
71 cm
0.10 mL
PTFE straight bore stopcock
PTFE
Blue print scale
PTFE
Buret
Not Autoclavable
Clear
25 mL
Acrylic
Used in quantitative chemical analysis to measure the volume of a liquid or a gas.
0.1 mL
0.1 mL
Key
0.1 mL
Class B
Videos
SDS
Documents

Documents

Promotions

Promotions