missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Class B Clear Glass Threaded Vials with Closures Attached
General-purpose vial
$89.86 - $161.45
Specifications
Classification | B |
---|---|
Material (Liner) | Polyvinyl |
Color | Clear |
Material | Borosilicate Glass |
Sterility | Non-sterile |
Includes: Attached black phenolic screw-top closures with polyvinyl-faced pulp liner
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Volume (English) | Volume (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Volume (English) | Volume (Metric) | Price | Quantity | |||
03-338E
|
N/A | 28 mm | 4 dr. | 14.8 mL |
Pack of 72 Each for $100.48
Case of 10 Pack for $1,004.80
|
|
|||
03-338A
|
N/A | 15 mm | 1 dr. | 3.7 mL |
Pack of 144 Each for $104.72
Case of 16 Pack for $1,675.52
|
|
|||
03-338AA
|
N/A | 12 mm | 0.5 dr. | 1.8 mL |
Pack of 144 Each for $145.00
Case of 36 Pack for $5,220.00
|
|
|||
03-338B
|
N/A | 16 mm | 1.5 dr. | 5.5 mL |
Pack of 144 Each for $130.86
Case of 18 Pack for $2,355.48
|
|
|||
03-338D
|
N/A | 19 mm | 3 dr. | 11.1 mL |
Pack of 144 Each for $139.97
Case of 12 Pack for $1,679.64
|
|
|||
03-338C
|
N/A | 17 mm | 2 dr. | 7.4 mL |
Pack of 144 Each for $122.72
Case of 12 Pack for $1,472.64
|
|
|||
03-338G
|
N/A | 28 mm | 6 dr. | 22.2 mL |
Pack of 72 Each for $103.14
Case of 8 Pack for $825.12
|
|
|||
03-338H
|
N/A | 23 mm | 6 dr. | 22.2 mL |
Pack of 144 Each for $161.45
Case of 6 Pack for $968.70
|
|
|||
03-338F
|
N/A | 21 mm | 4 dr. | 14.8 mL |
Pack of 144 Each for $151.14
Case of 8 Pack for $1,209.12
|
|
|||
03-338J
|
N/A | 25 mm | 8 dr. | 29.6 mL |
Pack of 72 Each for $89.86
Case of 8 Pack for $718.88
|
|
|||
03-338K
|
N/A | 28 mm | 9.5 dr. | 35.1 mL |
Pack of 72 Each for $107.69
Case of 6 Pack for $646.14
|
|
|||
03-338L
|
N/A | 28 mm | 11 dr. | 40.7 mL |
Pack of 72 Each for $119.10
Case of 6 Pack for $714.60
|
|
|||
Description
Vials for general-purpose use.
- Made from Type I, Class B borosilicate glass
- Attached black phenolic screw-top closures with polyvinyl-faced pulp liner
- GPI thread finish
Specifications
B | |
Clear | |
Non-sterile | |
For general purpose | |
Screw Cap |
Polyvinyl | |
Borosilicate Glass | |
Yes | |
Phenolic | |
Vial |