missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep
$101.21 - $367.56
Chemical Identifiers
CAS | 7439-95-4 |
---|---|
Molecular Formula | Mg |
Molecular Weight (g/mol) | 24.30 |
MDL Number | MFCD00085308 |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
PubChem CID | 5462224 |
ChEBI | CHEBI:25107 |
IUPAC Name | magnesium |
SMILES | [Mg] |
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mg | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
CHEBI:25107 | |
[Mg] |
Mg | |
MFCD00085308 | |
5462224 | |
magnesium |
Specifications
7439-95-4,7697-37-2,7732-18-5 | |
30 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00085308 | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
2% HNO3 |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Mg | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
Assurance | |
Magnesium Standard |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient