missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep

Supplier:  SPEX CertiPrep Group LLC PLMG22M

 View more versions of this product

Catalog No. PLMG22M



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mg1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Magnesium Standard
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Mg
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
Assurance
7439-95-4,7697-37-2,7732-18-5
30 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
MFCD00085308
[Mg]
24.30
CHEBI:25107
2% HNO3

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
CHEBI:25107
[Mg]
Mg
MFCD00085308
5462224
magnesium
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions