accessibility menu, dialog, popup

UserName

CAS RN 1257063-35-6

OOOONNNOOH

CAS RN 1257063-35-6

IUPAC Name: 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid
Synonyms: 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid
Molecular Weight (g/mol): 291.3
Molecular Formula: C11H21N3O6
InChi Key: BODPHGOBXPGJKO-UHFFFAOYSA-N
SMILES: OC(=O)CCOCCOCCOCCOCCN=[N+]=[N-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)