CAS RN 497-19-8
CAS RN 497-19-8
Sodium Carbonate Anhydrous (Powder/Certified ACS), Fisher Chemical™
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium Carbonate Anhydrous (Powder/HPLC), Fisher Chemical
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate solution, Honeywell Fluka™
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, Honeywell Fluka™
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium;carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 99.6%, ACS reagent, anhydrous
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 99.999%, (trace metal basis)
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium;carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 99.95%, extra pure, anhydrous
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 99.5%, extra pure, anhydrous
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 0.05N Standardized Solution
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC Name: disodium carbonate SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, 98%
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Sodium carbonate, anhydrous, ACS, 99.5% min
CAS: 497-19-8 Molecular Formula: CNa2O3 Molecular Weight (g/mol): 105.99 MDL Number: MFCD00003494 InChI Key: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O