missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Carbonate Anhydrous (Powder/Certified ACS), Fisher Chemical™
$298.58 - $9838.08
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
IUPAC Name | disodium carbonate |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
S263-500
|
Thermo Fisher Scientific
FLS263500 |
500 g | Poly Bottle |
Each for $298.58
Case of 6 Each for $1,791.48
|
|
|||||
S263-1
|
Fisher Chemical
FLS2631 |
1 kg | Poly Bottle |
Each for $409.92
Case of 6 Each for $2,459.52
|
|
|||||
S263-3
|
Fisher Chemical
FLS2633 |
3 kg | Poly Bottle |
Each for $800.83
Case of 4 Each for $3,203.32
|
|
|||||
S263-10
|
Thermo Fisher Scientific
FLS26310 |
10 kg | Poly Pail |
Each for $2,368.42
|
|
|||||
S26350
|
Fisher Chemical
FLS26350 |
50 kg | Fiber Drum |
Each for $9,838.08
|
|
|||||
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium carbonate |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
Specifications
1600°C | |
497-19-8 | |
11.3 | |
500 g | |
CNa2O3 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
≥99.5% | |
Pass Test | |
2.53g/cm³ | |
(dried basis)≥99.5% |
854°C | |
White | |
Solid | |
≥99.5% | |
MFCD00003494 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
Certified ACS | |
Poly Bottle | |
Sodium Carbonate Anhydrous |
Safety and Handling

WARNING!
Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:2
Flammability:0
Instability:1