accessibility menu, dialog, popup

UserName

CAS RN 528-92-7

CH3H3CH2COHNNH2O

CAS RN 528-92-7

IUPAC Name: [2-(propan-2-yl)pent-4-enoyl]urea
Synonyms: apronal
Molecular Weight (g/mol): 184.24
Molecular Formula: C9H16N2O2
InChi Key: KSUUMAWCGDNLFK-UHFFFAOYNA-N
SMILES: CC(C)C(CC=C)C(=O)NC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results