Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 147540050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,1,1,3,3,3-Hexafluoro-2-propanol | |
99.5 | |
-4.0°C | |
1.5960g/mL | |
>100°C | |
99.5% min. (GC) | |
C3H2F6O | |
(CF3)2CHOH | |
5 g | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
13529 | |
CHEBI:63104 | |
102mmHg at 20°C | |
Pure |
920-66-1 | |
100.0 | |
Colorless | |
59.0°C | |
Authentic | |
Glass bottle | |
1.2740 to 1.2770 | |
MFCD00011651 | |
1.596 | |
Solubility in water: 1000g/L (25°C). | |
C(C(F)(F)F)(C(F)(F)F)O | |
168.04 | |
1.65 mPa.s (20°C) | |
168.04 | |
99.5+% | |
Liquid |
Chemical Identifiers
920-66-1 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
13529 | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
CHEBI:63104 | |
C(C(F)(F)F)(C(F)(F)F)O |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 213-059-4
RUO – Research Use Only