Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure, Thermo Scientific Chemicals
$86.56 - $1806.44
Chemical Identifiers
CAS | 920-66-1 |
---|---|
MDL Number | MFCD00011651 |
InChI Key | BYEAHWXPCBROCE-UHFFFAOYSA-N |
Synonym | 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol |
PubChem CID | 13529 |
ChEBI | CHEBI:63104 |
IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-ol |
SMILES | C(C(F)(F)F)(C(F)(F)F)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147540050
|
Thermo Scientific Chemicals
147540050 |
5 g | Glass bottle |
Each for $86.56
|
|
||||
AC147541000
|
Thermo Scientific Chemicals
147541000 |
100 g | Glass Bottle |
Each for $597.38
|
|
||||
AC147545000
|
Thermo Scientific Chemicals
147545000 |
500 g | Glass bottle |
Each for $1,806.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
920-66-1 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
13529 | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
CHEBI:63104 | |
C(C(F)(F)F)(C(F)(F)F)O |
Specifications
920-66-1 | |
100.0 | |
Colorless | |
59.0°C | |
Authentic | |
Glass bottle | |
1.2740 to 1.2770 | |
MFCD00011651 | |
1.596 | |
Solubility in water: 1000g/L (25°C). | |
C(C(F)(F)F)(C(F)(F)F)O | |
168.04 | |
1.65 mPa.s (20°C) | |
168.04 | |
99.5+% | |
Liquid |
99.5 | |
-4.0°C | |
1.5960g/mL | |
>100°C | |
99.5% min. (GC) | |
C3H2F6O | |
(CF3)2CHOH | |
5 g | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
13529 | |
CHEBI:63104 | |
102mmHg at 20°C | |
Pure | |
1,1,1,3,3,3-Hexafluoro-2-propanol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 213-059-4
RUO – Research Use Only