missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,1-Trifluoroacetone cyanohydrin, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1131806

 View more versions of this product

Catalog No. AAL1131806


Explore available promotions

Description

Description

A chlorinated acetone derivative for organic synthesis and proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chlorinated acetone derivative for organic synthesis and proteomics research

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Specifications

Specifications

1,1,1-Trifluoroacetone cyanohydrin
1.276
60°C (140°F)
1.338
5 g
1758723
1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol
XDCMNDCKYSQKAX-UHFFFAOYSA-N
3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
274400
95%
335-08-0
68°C to 69°C (40 mmHg)
C4H4F3NO
MFCD00040885
UN2929
Moisture sensitive
Fully miscible in water.
CC(C#N)(C(F)(F)F)O
139.077
139.08

Chemical Identifiers

335-08-0
139.077
XDCMNDCKYSQKAX-UHFFFAOYSA-N
274400
CC(C#N)(C(F)(F)F)O
C4H4F3NO
MFCD00040885
1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol
3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only