Learn More
1,1,1-Trifluoroacetone cyanohydrin, 95%
CAS: 335-08-0 | C4H4F3NO | 139.077 g/mol
$280.68 - $1128.03
Chemical Identifiers
| CAS | 335-08-0 |
|---|---|
| Molecular Formula | C4H4F3NO |
| Molecular Weight (g/mol) | 139.077 |
| MDL Number | MFCD00040885 |
| InChI Key | XDCMNDCKYSQKAX-UHFFFAOYSA-N |
| Synonym | 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol |
| PubChem CID | 274400 |
| IUPAC Name | 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile |
| SMILES | CC(C#N)(C(F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1131806
|
Thermo Scientific Chemicals
L1131806 |
5 g |
Each for $280.68
|
|
|||||
|
AAL1131814
|
Thermo Scientific Chemicals
L1131814 |
25 g |
Each for $1,128.03
|
|
|||||
Description
A chlorinated acetone derivative for organic synthesis and proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA chlorinated acetone derivative for organic synthesis and proteomics research
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Chemical Identifiers
| 335-08-0 | |
| 139.077 | |
| XDCMNDCKYSQKAX-UHFFFAOYSA-N | |
| 274400 | |
| CC(C#N)(C(F)(F)F)O |
| C4H4F3NO | |
| MFCD00040885 | |
| 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol | |
| 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile |
Specifications
| 335-08-0 | |
| 68°C to 69°C (40 mmHg) | |
| C4H4F3NO | |
| MFCD00040885 | |
| UN2929 | |
| Moisture sensitive | |
| Fully miscible in water. | |
| CC(C#N)(C(F)(F)F)O | |
| 139.077 | |
| 139.08 | |
| 1,1,1-Trifluoroacetone cyanohydrin |
| 1.276 | |
| 60°C (140°F) | |
| 1.338 | |
| 5 g | |
| 1758723 | |
| 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol | |
| XDCMNDCKYSQKAX-UHFFFAOYSA-N | |
| 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile | |
| 274400 | |
| 95% |
Safety and Handling
GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
H226-H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only