missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2-Trichloroethane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 1369432500

 View more versions of this product

Catalog No. AC139432500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 1, 2-Trichloroethane
-37.0°C
1.4300g/mL
97.5% min. (GC)
C2H3Cl3
ClCH2CHCl2
250 mL
1.43
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
UBOXGVDOUJQMTN-UHFFFAOYSA-N
1,1,2-trichloroethane
6574
CHEBI:36018
98%
79-00-5
Colorless
110.0°C to 115.0°C
Glass bottle
1.4700 to 1.4720
MFCD00000852
01,85
15,9802
Solubility in water: 4g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,miscible with alcohols,ethers,esters and ketones
ClCC(Cl)Cl
133.40
1.69 mPa.s (25°C)
133.4
Liquid

Chemical Identifiers

79-00-5
133.40
UBOXGVDOUJQMTN-UHFFFAOYSA-N
6574
1,1,2-trichloroethane
C2H3Cl3
MFCD00000852
ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
CHEBI:36018
ClCC(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 201-166-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only