missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2-Trichloroethane, 98%
CAS: 79-00-5 | C2H3Cl3 | 133.40 g/mol
$167.03 - $167.03
Chemical Identifiers
| CAS | 79-00-5 |
|---|---|
| Molecular Formula | C2H3Cl3 |
| Molecular Weight (g/mol) | 133.40 |
| MDL Number | MFCD00000852 |
| InChI Key | UBOXGVDOUJQMTN-UHFFFAOYSA-N |
| Synonym | ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t |
| PubChem CID | 6574 |
| ChEBI | CHEBI:36018 |
| IUPAC Name | 1,1,2-trichloroethane |
| SMILES | ClCC(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC139432500
|
Thermo Scientific Chemicals
1369432500 |
250 mL | Glass bottle |
Each for $167.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 79-00-5 | |
| 133.40 | |
| UBOXGVDOUJQMTN-UHFFFAOYSA-N | |
| 6574 | |
| 1,1,2-trichloroethane |
| C2H3Cl3 | |
| MFCD00000852 | |
| ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t | |
| CHEBI:36018 | |
| ClCC(Cl)Cl |
Specifications
| 79-00-5 | |
| Colorless | |
| 110.0°C to 115.0°C | |
| Glass bottle | |
| 1.4700 to 1.4720 | |
| MFCD00000852 | |
| 01,85 | |
| 15,9802 | |
| Solubility in water: 4g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,miscible with alcohols,ethers,esters and ketones | |
| ClCC(Cl)Cl | |
| 133.40 | |
| 1.69 mPa.s (25°C) | |
| 133.4 | |
| Liquid |
| -37.0°C | |
| 1.4300g/mL | |
| 97.5% min. (GC) | |
| C2H3Cl3 | |
| ClCH2CHCl2 | |
| 250 mL | |
| 1.43 | |
| ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t | |
| UBOXGVDOUJQMTN-UHFFFAOYSA-N | |
| 1,1,2-trichloroethane | |
| 6574 | |
| CHEBI:36018 | |
| 98% | |
| 1, 1, 2-Trichloroethane |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 201-166-9
RUO – Research Use Only