missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Dibromoethane, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 406480250

 View more versions of this product

Catalog No. AC406480250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,1-Dibromoethane
557-91-5
100.0
2.0600g/mL
Authentic
Glass bottle
1.5090 to 1.5120
MFCD00041719
2.06
APQIUTYORBAGEZ-UHFFFAOYSA-N
1,1-dibromoethane
potassium carbonate
187.86
Liquid
99%
98.5
Colorless to Brown
107.0°C
98.5% min. (GC)
C2H4Br2
CH3CHBr2
25 g
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
CC(Br)Br
187.86
11201
99%

Chemical Identifiers

557-91-5
187.86
APQIUTYORBAGEZ-UHFFFAOYSA-N
11201
CC(Br)Br
C2H4Br2
MFCD00041719
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
1,1-dibromoethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-184-9

RTECSNumber : KH9000000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only