missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1-Dibromoethane, 99%, stabilized, Thermo Scientific Chemicals

$322.01 - $322.01

Chemical Identifiers

CAS 557-91-5
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.86
MDL Number MFCD00041719
InChI Key APQIUTYORBAGEZ-UHFFFAOYSA-N
Synonym ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
PubChem CID 11201
IUPAC Name 1,1-dibromoethane
SMILES CC(Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC406480250
SDS View Documents
Thermo Scientific Chemicals
406480250
25 g Glass bottle
Each for $322.01
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

557-91-5
187.86
APQIUTYORBAGEZ-UHFFFAOYSA-N
11201
CC(Br)Br
C2H4Br2
MFCD00041719
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
1,1-dibromoethane

Specifications

557-91-5
100.0
2.0600g/mL
Authentic
Glass bottle
1.5090 to 1.5120
MFCD00041719
2.06
APQIUTYORBAGEZ-UHFFFAOYSA-N
1,1-dibromoethane
potassium carbonate
187.86
Liquid
98.5
Colorless to Brown
107.0°C
98.5% min. (GC)
C2H4Br2
CH3CHBr2
25 g
ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
CC(Br)Br
187.86
11201
99%
1,1-Dibromoethane, 99%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-184-9

RTECSNumber : KH9000000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only