missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Diaminodecane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 112130050

 View more versions of this product

Catalog No. AC112130050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 10-Diaminodecane
646-25-3
100.0
White to Yellow
Authentic
Glass bottle
H2N(CH2)10NH2
5 g
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
C(CCCCCN)CCCCN
172.31
172.31
Chunks and/or Powder
97%
96.0
62.0°C to 63.0°C
140.0°C (12.0 mmHg)
96% min. (GC)
C10H24N2
MFCD00008151
04, 273
YQLZOAVZWJBZSY-UHFFFAOYSA-N
decane-1,10-diamine
1317
97%

Chemical Identifiers

646-25-3
172.31
YQLZOAVZWJBZSY-UHFFFAOYSA-N
1317
C(CCCCCN)CCCCN
C10H24N2
MFCD00008151
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
decane-1,10-diamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 211-471-9

RTECSNumber : HD7175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only