missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Diaminodecane, 97%, Thermo Scientific Chemicals

$89.45 - $270.66

Chemical Identifiers

CAS 646-25-3
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.31
MDL Number MFCD00008151
InChI Key YQLZOAVZWJBZSY-UHFFFAOYSA-N
Synonym 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
PubChem CID 1317
IUPAC Name decane-1,10-diamine
SMILES C(CCCCCN)CCCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112130050
SDS View Documents
Thermo Scientific Chemicals
112130050
5 g Glass bottle
Each for $89.45
 
AC112130250
SDS View Documents
Thermo Scientific Chemicals
112130250
25 g Glass bottle
Each for $270.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

646-25-3
172.31
YQLZOAVZWJBZSY-UHFFFAOYSA-N
1317
C(CCCCCN)CCCCN
C10H24N2
MFCD00008151
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
decane-1,10-diamine

Specifications

646-25-3
100.0
White to Yellow
Authentic
Glass bottle
H2N(CH2)10NH2
5 g
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
C(CCCCCN)CCCCN
172.31
172.31
Chunks and/or Powder
96.0
62.0°C to 63.0°C
140.0°C (12.0 mmHg)
96% min. (GC)
C10H24N2
MFCD00008151
04, 273
YQLZOAVZWJBZSY-UHFFFAOYSA-N
decane-1,10-diamine
1317
97%
1, 10-Diaminodecane, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 211-471-9

RTECSNumber : HD7175000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only