Learn More
1,10-Diaminodecane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 112130250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 10-Diaminodecane | |
646-25-3 | |
100.0 | |
White to Yellow | |
Authentic | |
Glass bottle | |
H2N(CH2)10NH2 | |
25 g | |
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
C(CCCCCN)CCCCN | |
172.31 | |
172.31 | |
Chunks and/or Powder |
97% | |
96.0 | |
62.0°C to 63.0°C | |
140.0°C (12.0 mmHg) | |
96% min. (GC) | |
C10H24N2 | |
MFCD00008151 | |
04, 273 | |
YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
decane-1,10-diamine | |
1317 | |
97% |
Chemical Identifiers
646-25-3 | |
172.31 | |
YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
1317 | |
C(CCCCCN)CCCCN |
C10H24N2 | |
MFCD00008151 | |
1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
decane-1,10-diamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 211-471-9
RTECSNumber : HD7175000
TSCA : TSCA
RUO – Research Use Only