missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 417120010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
1, 10-Phenanthroline monohydrate | |
97°C to 101°C | |
Authentic | |
C12H10N2O | |
1 g | |
15, 7328 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,acetone and benzene | |
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
198.23 | |
198.23 | |
Needle-like Crystals |
5144-89-8,66-71-7 | |
White | |
Glass bottle | |
MFCD00149973 | |
23, 227 | |
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate | |
PPQJCISYYXZCAE-UHFFFAOYSA-N | |
1,10-phenanthroline;hydrate | |
21226 | |
ACS Reagent |
Chemical Identifiers
5144-89-8 | |
198.23 | |
PPQJCISYYXZCAE-UHFFFAOYSA-N | |
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
C12H10N2O | |
MFCD00149973 | |
21226 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
GHS Signal Word: Danger
RTECSNumber : SF8437000
TSCA : TSCA
RUO – Research Use Only