missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals

$50.00 - $1105.15

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC417120010
SDS View Documents
Thermo Scientific Chemicals
417120010
1 g Glass bottle
Each for $50.00
Case of 6 Each for $0.00
 
AC417120050
SDS View Documents
Thermo Scientific Chemicals
417120050
5 g Glass bottle
Each for $102.53
Case of 6 Each for $0.00
 
AC417120250
SDS View Documents
Thermo Scientific Chemicals
417120250
25 g Glass bottle
Each for $363.40
 
AC417121000
SDS View Documents
Thermo Scientific Chemicals
417121000
100 g Glass bottle
Each for $1,105.15
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

5144-89-8
198.23
PPQJCISYYXZCAE-UHFFFAOYSA-N
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
C12H10N2O
MFCD00149973
21226

Specifications

5144-89-8,66-71-7
White
Glass bottle
MFCD00149973
23, 227
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline;hydrate
21226
ACS Reagent
1, 10-Phenanthroline monohydrate
97°C to 101°C
Authentic
C12H10N2O
1 g
15, 7328
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,acetone and benzene
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
198.23
198.23
Needle-like Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

GHS Signal Word: Danger

RTECSNumber : SF8437000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only