missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,10-Phenanthroline monohydrate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 417120250

 View more versions of this product

Catalog No. AC417120250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

1, 10-Phenanthroline monohydrate
97°C to 101°C
Authentic
C12H10N2O
25 g
15, 7328
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,acetone and benzene
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
198.23
198.23
Needle-like Crystals
5144-89-8,66-71-7
White
Glass bottle
MFCD00149973
23, 227
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate
21226
ACS Reagent

Chemical Identifiers

5144-89-8
198.23
PPQJCISYYXZCAE-UHFFFAOYSA-N
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
C12H10N2O
MFCD00149973
21226
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

GHS Signal Word: Danger

RTECSNumber : SF8437000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only