missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 348070250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2,3,4-Tetramethyl-1,3-cyclopentadiene | |
Yellow | |
142.0°C | |
Authentic | |
Glass bottle | |
1.4700 to 1.4760 | |
0.808 | |
VNPQQEYMXYCAEZ-UHFFFAOYSA-N | |
cyclopentanecarbonitrile | |
77935 | |
85% |
4249-10-9 | |
0.8080g/mL | |
42°C | |
80% min. (GC) | |
C9H14 | |
25 g | |
cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile | |
CC1=C(C)C(C)=C(C)C1 | |
122.21 | |
122.21 | |
Liquid |
Chemical Identifiers
4249-10-9 | |
VNPQQEYMXYCAEZ-UHFFFAOYSA-N | |
77935 |
122.21 | |
cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile | |
CC1=C(C)C(C)=C(C)C1 |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only