Learn More
1,2,3,4-Tetramethylbenzene, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1975614
Description
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
1,2,3,4-Tetramethylbenzene | |
0.9 | |
68°C (154°F) | |
1.519 | |
25 g | |
prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene | |
UOHMMEJUHBCKEE-UHFFFAOYSA-N | |
1,2,3,4-tetramethylbenzene | |
10263 | |
134.22 |
488-23-3 | |
203°C | |
C10H14 | |
MFCD00008521 | |
1904390 | |
Immiscible with water. | |
CC1=C(C(=C(C=C1)C)C)C | |
134.222 | |
CHEBI:38997 | |
95% |
Chemical Identifiers
488-23-3 | |
134.222 | |
UOHMMEJUHBCKEE-UHFFFAOYSA-N | |
10263 | |
1,2,3,4-tetramethylbenzene |
C10H14 | |
MFCD00008521 | |
prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene | |
CHEBI:38997 | |
CC1=C(C(=C(C=C1)C)C)C |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 207-673-1
RTECSNumber : DC0465000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only