missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3,4-Tetramethylbenzene, 95%, Thermo Scientific Chemicals

$230.10 - $938.66

Chemical Identifiers

CAS 488-23-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008521
InChI Key UOHMMEJUHBCKEE-UHFFFAOYSA-N
Synonym prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
PubChem CID 10263
ChEBI CHEBI:38997
IUPAC Name 1,2,3,4-tetramethylbenzene
SMILES CC1=C(C(=C(C=C1)C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1975606
View Documents
Thermo Scientific Chemicals
L1975606
5 g
Each for $230.10
 
AAL1975614
View Documents
Thermo Scientific Chemicals
L1975614
25 g
Each for $938.66
 
Description

Description

1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetramethylbenzene is used as a starting material in the preparation of 1,2,34-tetramethyl benzene-5-chlorosulfonyl chloride by reacting with chlorosulfonic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

488-23-3
134.222
UOHMMEJUHBCKEE-UHFFFAOYSA-N
10263
1,2,3,4-tetramethylbenzene
C10H14
MFCD00008521
prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
CHEBI:38997
CC1=C(C(=C(C=C1)C)C)C

Specifications

488-23-3
203°C
C10H14
MFCD00008521
1904390
Immiscible with water.
CC1=C(C(=C(C=C1)C)C)C
134.222
CHEBI:38997
95%
0.9
68°C (154°F)
1.519
5 g
prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
UOHMMEJUHBCKEE-UHFFFAOYSA-N
1,2,3,4-tetramethylbenzene
10263
134.22
1,2,3,4-Tetramethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 207-673-1

RTECSNumber : DC0465000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only