missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6681806

 View more versions of this product

Catalog No. AAJ6681806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
65°C to 69°C
C11H16O7
5 g
Soluble in ethanol
CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
260.24
260.24
Powder
62211-93-2
Yellow to Brown
MFCD08458459,MFCD08458459
1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
NXEJETQVUQAKTO-UHFFFAOYNA-N
[(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
70847275
98%

Chemical Identifiers

62211-93-2
260.24
NXEJETQVUQAKTO-UHFFFAOYNA-N
70847275
CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
C11H16O7
MFCD08458459,MFCD08458459
1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
[(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only