missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

$70.86 - $195.52

Chemical Identifiers

CAS 62211-93-2
Molecular Formula C11H16O7
Molecular Weight (g/mol) 260.24
MDL Number MFCD08458459,MFCD08458459
InChI Key NXEJETQVUQAKTO-UHFFFAOYNA-N
Synonym 1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
PubChem CID 70847275
IUPAC Name [(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
SMILES CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6681803
View Documents
Thermo Scientific Chemicals
J6681803
1 g
Each for $70.86
 
AAJ6681806
View Documents
Thermo Scientific Chemicals
J6681806
5 g
Each for $195.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62211-93-2
260.24
NXEJETQVUQAKTO-UHFFFAOYNA-N
70847275
CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
C11H16O7
MFCD08458459,MFCD08458459
1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
[(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate

Specifications

62211-93-2
Yellow to Brown
MFCD08458459,MFCD08458459
1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
NXEJETQVUQAKTO-UHFFFAOYNA-N
[(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
70847275
98%
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
65°C to 69°C
C11H16O7
1 g
Soluble in ethanol
CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
260.24
260.24
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only