missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2,3-Trichloropropane 98.0+%, TCI America™
$27.49 - $27.49
Chemical Identifiers
CAS | 96-18-4 |
---|---|
Molecular Formula | C3H5Cl3 |
Molecular Weight (g/mol) | 147.42 |
MDL Number | MFCD00000946 |
InChI Key | CFXQEHVMCRXUSD-UHFFFAOYSA-N |
Synonym | trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132 |
PubChem CID | 7285 |
ChEBI | CHEBI:34036 |
IUPAC Name | 1,2,3-trichloropropane |
SMILES | ClCC(Cl)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T039525G
|
TCI America
T039525G |
25 g |
Each for $27.49
|
|
|||||
Chemical Identifiers
96-18-4 | |
147.42 | |
CFXQEHVMCRXUSD-UHFFFAOYSA-N | |
7285 | |
1,2,3-trichloropropane |
C3H5Cl3 | |
MFCD00000946 | |
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132 | |
CHEBI:34036 | |
ClCC(Cl)CCl |
Specifications
96-18-4 | |
156°C | |
MFCD00000946 | |
2810 | |
CFXQEHVMCRXUSD-UHFFFAOYSA-N | |
1,2,3-trichloropropane | |
7285 | |
147.42 | |
Liquid |
Yellow | |
C3H5Cl3 | |
25 g | |
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132 | |
ClCC(Cl)CCl | |
147.42 | |
CHEBI:34036 | |
≥98.0% (GC) | |
1,2,3-Trichloropropane |
Safety and Handling
EINECSNumber : (2)-0083
RTECSNumber : TZ9275000
TSCA : Yes