missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2,3-Trichloropropane 98.0+%, TCI America™

Supplier:  TCI America T039525G

 View more versions of this product

Catalog No. T039525G


Explore available promotions

Description

Description

Specifications

Specifications

1,2,3-Trichloropropane
Yellow
C3H5Cl3
25 g
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
ClCC(Cl)CCl
147.42
CHEBI:34036
≥98.0% (GC)
96-18-4
156°C
MFCD00000946
2810
CFXQEHVMCRXUSD-UHFFFAOYSA-N
1,2,3-trichloropropane
7285
147.42
Liquid

Chemical Identifiers

96-18-4
147.42
CFXQEHVMCRXUSD-UHFFFAOYSA-N
7285
1,2,3-trichloropropane
C3H5Cl3
MFCD00000946
trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
CHEBI:34036
ClCC(Cl)CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0083

RTECSNumber : TZ9275000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions