missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2:5,6-Di-O-isopropylidene-D-mannitol, Thermo Scientific Chemicals
$76.52 - $290.36
Chemical Identifiers
CAS | 1707-77-3 |
---|---|
Molecular Formula | C12H22O6 |
Molecular Weight (g/mol) | 262.302 |
MDL Number | MFCD00003211 |
InChI Key | ODYBCPSCYHAGHA-ZYUZMQFOSA-N |
Synonym | 1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213 |
PubChem CID | 96011 |
IUPAC Name | (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol |
SMILES | CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1212706
|
Thermo Scientific Chemicals
L1212706 |
5 g |
Each for $76.52
|
|
|||||
AAL1212714
|
Thermo Scientific Chemicals
L1212714 |
25 g |
Each for $290.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1707-77-3 | |
262.302 | |
ODYBCPSCYHAGHA-ZYUZMQFOSA-N | |
96011 | |
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C |
C12H22O6 | |
MFCD00003211 | |
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213 | |
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol |
Specifications
1707-77-3 | |
C12H22O6 | |
5 g | |
1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213 | |
+6° (c=5 in Chloroform) | |
(1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol | |
96011 | |
1,2:5,6-Di-O-isopropylidene-D-mannitol |
120°C to 122°C | |
MFCD00003211 | |
83765 | |
ODYBCPSCYHAGHA-ZYUZMQFOSA-N | |
CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C | |
262.302 | |
262.31 |
Safety and Handling
EINECSNumber : 216-954-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only