missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 391690050

 View more versions of this product

Catalog No. AC391690050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 2-Bis(2-iodoethoxy)ethane
96.0
2.0280g/mL
Authentic
Glass bottle
I(CH2)2O(CH2)2O(CH2)2I
5g
1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
C(COCCI)OCCI
369.96
593436
97%
36839-55-1
100.0
113°C
96% min. (GC)
C6H12I2O2
MFCD00075008
2.028
BCAGFJXMCZSAHD-UHFFFAOYSA-N
1,2-bis(2-iodoethoxy)ethane
copper
369.96

Chemical Identifiers

36839-55-1
369.96
BCAGFJXMCZSAHD-UHFFFAOYSA-N
593436
C(COCCI)OCCI
C6H12I2O2
MFCD00075008
1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
1,2-bis(2-iodoethoxy)ethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only