missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized, Thermo Scientific™

Chemical Identifiers

CAS 36839-55-1
Molecular Formula C6H12I2O2
Molecular Weight (g/mol) 369.96
MDL Number MFCD00075008
InChI Key BCAGFJXMCZSAHD-UHFFFAOYSA-N
Synonym 1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
PubChem CID 593436
IUPAC Name 1,2-bis(2-iodoethoxy)ethane
SMILES C(COCCI)OCCI
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391690050
SDS View Documents
Thermo Scientific Chemicals
391690050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC391690250
SDS View Documents
Thermo Scientific Chemicals
391690250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

36839-55-1
369.96
BCAGFJXMCZSAHD-UHFFFAOYSA-N
593436
C(COCCI)OCCI
C6H12I2O2
MFCD00075008
1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
1,2-bis(2-iodoethoxy)ethane

Specifications

36839-55-1
100.0
113°C
96% min. (GC)
C6H12I2O2
MFCD00075008
2.028
BCAGFJXMCZSAHD-UHFFFAOYSA-N
1,2-bis(2-iodoethoxy)ethane
copper
369.96
1, 2-Bis(2-iodoethoxy)ethane
96.0
2.0280g/mL
Authentic
Glass bottle
I(CH2)2O(CH2)2O(CH2)2I
5g
1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
C(COCCI)OCCI
369.96
593436
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only