missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H60592MD

 View more versions of this product

Catalog No. AAH60592MD


Explore available promotions

Description

Description

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
Specifications

Specifications

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene
66°C to 76°C
MFCD00142336
4810602
r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos
AJNZWRKTWQLAJK-KLHDSHLOSA-N
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane
2734551
≥97%
147253-67-6
C18H28P2
250 mg
Air Sensitive
Insoluble in water.
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C
306.37
306.36

Chemical Identifiers

147253-67-6
306.37
AJNZWRKTWQLAJK-KLHDSHLOSA-N
2734551
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C
C18H28P2
MFCD00142336
r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only