missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%, Thermo Scientific Chemicals
$253.84 - $851.00
Chemical Identifiers
CAS | 147253-67-6 |
---|---|
Molecular Formula | C18H28P2 |
Molecular Weight (g/mol) | 306.37 |
MDL Number | MFCD00142336 |
InChI Key | AJNZWRKTWQLAJK-KLHDSHLOSA-N |
Synonym | r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos |
PubChem CID | 2734551 |
IUPAC Name | (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane |
SMILES | CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH60592MD
|
Thermo Scientific Chemicals
H60592MD |
250 mg |
Each for $253.84
|
|
||||||
AAH6059203
|
Thermo Scientific Chemicals
H6059203 |
1 g |
Each for $851.00
|
|
||||||
Description
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
Chemical Identifiers
147253-67-6 | |
306.37 | |
AJNZWRKTWQLAJK-KLHDSHLOSA-N | |
2734551 | |
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C |
C18H28P2 | |
MFCD00142336 | |
r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos | |
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane |
Specifications
147253-67-6 | |
C18H28P2 | |
250 mg | |
Air Sensitive | |
Insoluble in water. | |
CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C | |
306.37 | |
306.36 | |
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene |
66°C to 76°C | |
MFCD00142336 | |
4810602 | |
r,r-me-duphos, methyl-duphos, r,r, r,r-methyl-duphos, unii-h5w03d1haq, h5w03d1haq, --1,2-bis 2r,5r-2,5-dimethylphospholano benzene, r,r-meduphos, methyl-duphos,-, 2r,5r-me-duphos | |
AJNZWRKTWQLAJK-KLHDSHLOSA-N | |
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane | |
2734551 | |
≥97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only