Learn More
1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 321470010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Bis(dichlorophosphino)ethane | |
28240-69-9 | |
100.0 | |
70.0°C (1.0 mmHg) | |
Glass bottle | |
1.5875 to 1.5935 | |
MFCD00009609 | |
1.53 | |
Solubility in water: reacts | |
C(CP(Cl)Cl)P(Cl)Cl | |
231.81 | |
231.81 |
97% | |
96.0 | |
1.5300g/mL | |
97% | |
C2H4Cl4P2 | |
Cl2PCH2CH2PCl2 | |
1 g | |
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472 | |
SBWAJHLQMFBNIN-UHFFFAOYSA-N | |
dichloro(2-dichlorophosphanylethyl)phosphane | |
119904 | |
97% |
Chemical Identifiers
28240-69-9 | |
231.81 | |
SBWAJHLQMFBNIN-UHFFFAOYSA-N | |
119904 | |
C(CP(Cl)Cl)P(Cl)Cl |
C2H4Cl4P2 | |
MFCD00009609 | |
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472 | |
dichloro(2-dichlorophosphanylethyl)phosphane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
TSCA : TSCA
RUO – Research Use Only