missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals

$135.35 - $434.18

Chemical Identifiers

CAS 28240-69-9
Molecular Formula C2H4Cl4P2
Molecular Weight (g/mol) 231.81
MDL Number MFCD00009609
InChI Key SBWAJHLQMFBNIN-UHFFFAOYSA-N
Synonym 1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
PubChem CID 119904
IUPAC Name dichloro(2-dichlorophosphanylethyl)phosphane
SMILES C(CP(Cl)Cl)P(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC321470010
SDS View Documents
Thermo Scientific Chemicals
321470010
1 g Glass bottle
Each for $135.35
 
AC321470050
SDS View Documents
Thermo Scientific Chemicals
321470050
5 g Glass bottle
Each for $434.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28240-69-9
231.81
SBWAJHLQMFBNIN-UHFFFAOYSA-N
119904
C(CP(Cl)Cl)P(Cl)Cl
C2H4Cl4P2
MFCD00009609
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
dichloro(2-dichlorophosphanylethyl)phosphane

Specifications

28240-69-9
100.0
70.0°C (1.0 mmHg)
Glass bottle
1.5875 to 1.5935
MFCD00009609
1.53
Solubility in water: reacts
C(CP(Cl)Cl)P(Cl)Cl
231.81
231.81
1, 2-Bis(dichlorophosphino)ethane, 97%
96.0
1.5300g/mL
97%
C2H4Cl4P2
Cl2PCH2CH2PCl2
1 g
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
SBWAJHLQMFBNIN-UHFFFAOYSA-N
dichloro(2-dichlorophosphanylethyl)phosphane
119904
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only