Learn More
1,2-Bis(dimethoxyphosphoryl)benzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03927309
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,2-Bis(dimethoxyphosphoryl)benzene | |
80°C to 82°C | |
MFCD00142461 | |
UN3464 | |
TUKTVDDATWNXSN-UHFFFAOYSA-N | |
1,2-bis(dimethoxyphosphoryl)benzene | |
2733153 | |
99% |
15104-46-8 | |
C10H16O6P2 | |
10 g | |
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one | |
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC | |
294.18 | |
294.18 | |
Crystalline |
Chemical Identifiers
15104-46-8 | |
294.18 | |
TUKTVDDATWNXSN-UHFFFAOYSA-N | |
2733153 | |
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC |
C10H16O6P2 | |
MFCD00142461 | |
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one | |
1,2-bis(dimethoxyphosphoryl)benzene |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only