missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Bis(dimethoxyphosphoryl)benzene, 99%, Thermo Scientific Chemicals

$184.96 - $798.18

Chemical Identifiers

CAS 15104-46-8
Molecular Formula C10H16O6P2
Molecular Weight (g/mol) 294.18
MDL Number MFCD00142461
InChI Key TUKTVDDATWNXSN-UHFFFAOYSA-N
Synonym 1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one
PubChem CID 2733153
IUPAC Name 1,2-bis(dimethoxyphosphoryl)benzene
SMILES COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3927304
View Documents
Thermo Scientific Chemicals
03927304
2 g
Each for $184.96
 
AA3927309
View Documents
Thermo Scientific Chemicals
03927309
10 g
Each for $798.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15104-46-8
294.18
TUKTVDDATWNXSN-UHFFFAOYSA-N
2733153
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC
C10H16O6P2
MFCD00142461
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one
1,2-bis(dimethoxyphosphoryl)benzene

Specifications

15104-46-8
C10H16O6P2
2 g
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC
294.18
294.18
Crystalline
80°C to 82°C
MFCD00142461
UN3464
TUKTVDDATWNXSN-UHFFFAOYSA-N
1,2-bis(dimethoxyphosphoryl)benzene
2733153
99%
1,2-Bis(dimethoxyphosphoryl)benzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only