Learn More
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, Thermo Scientific Chemicals
$166.93 - $166.93
Chemical Identifiers
CAS | 14647-23-5 |
---|---|
Molecular Formula | C26H24Cl2NiP2 |
Molecular Weight (g/mol) | 528.02 |
MDL Number | MFCD00013313 |
InChI Key | XXECWTBMGGXMKP-UHFFFAOYSA-L |
Synonym | 1,2-bis diphenylphosphino ethane nickel ii chloride |
PubChem CID | 131664338 |
IUPAC Name | nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride |
SMILES | Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC363230050
|
Thermo Scientific Chemicals
363230050 |
5 g | Glass bottle |
Each for $166.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14647-23-5 | |
528.02 | |
XXECWTBMGGXMKP-UHFFFAOYSA-L | |
131664338 | |
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C26H24Cl2NiP2 | |
MFCD00013313 | |
1,2-bis diphenylphosphino ethane nickel ii chloride | |
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride |
Specifications
14647-23-5 | |
100.0 | |
Orange to Red | |
Authentic | |
Glass bottle | |
MFCD00013313 | |
1,2-bis diphenylphosphino ethane nickel ii chloride | |
XXECWTBMGGXMKP-UHFFFAOYSA-L | |
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride | |
131664338 | |
99% | |
1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride |
98.5 | |
263.0°C to 265.0°C | |
98.5% min. | |
99% | |
C26H24Cl2NiP2 | |
5 g | |
Solubility in water: insoluble in water | |
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
528.02 | |
528.02 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only