missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Butylene Oxide 99.0+%, TCI America™

Supplier:  TCI America B071925ML

 View more versions of this product

Catalog No. B071925ML


Explore available promotions

Description

Description

Specifications

Specifications

1,2-Butylene Oxide
Colorless
C4H8O
25 mL
1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
CCC1CO1
72.107
CHEBI:82326
≥99.0% (GC)
106-88-7
63°C
MFCD00005153
3022
RBACIKXCRWGCBB-UHFFFAOYSA-N
2-ethyloxirane
7834
72.11
Liquid

Chemical Identifiers

106-88-7
72.107
RBACIKXCRWGCBB-UHFFFAOYSA-N
7834
2-ethyloxirane
C4H8O
MFCD00005153
1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
CHEBI:82326
CCC1CO1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0229

RTECSNumber : EK3675000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions