missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Butylene Oxide 99.0+%, TCI America™

$35.56 - $92.83

Chemical Identifiers

CAS 106-88-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00005153
InChI Key RBACIKXCRWGCBB-UHFFFAOYSA-N
Synonym 1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
PubChem CID 7834
ChEBI CHEBI:82326
IUPAC Name 2-ethyloxirane
SMILES CCC1CO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B071925ML
View Documents
TCI America
B071925ML
25 mL
Each for $35.56
 
B0719100ML
View Documents
TCI America
B0719100ML
100 mL
Each for $55.30
 
B0719500ML
View Documents
TCI America
B0719500ML
500 mL
Each for $92.83
 
Description

Description

Specifications

Chemical Identifiers

106-88-7
72.107
RBACIKXCRWGCBB-UHFFFAOYSA-N
7834
2-ethyloxirane
C4H8O
MFCD00005153
1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
CHEBI:82326
CCC1CO1

Specifications

106-88-7
63°C
MFCD00005153
3022
RBACIKXCRWGCBB-UHFFFAOYSA-N
2-ethyloxirane
7834
72.11
Liquid
Colorless
C4H8O
25 mL
1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
CCC1CO1
72.107
CHEBI:82326
≥99.0% (GC)
1,2-Butylene Oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0229

RTECSNumber : EK3675000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'