missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0703314

 View more versions of this product

Catalog No. AAL0703314



Description

Description

1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1,2-Epoxy-7-octene
0.85
38°C (100°F)
1.435
25 g
1305290
Not miscible in water.
C=CCCCCC1CO1
126.199
126.2
19600-63-6
167°C
C8H14O
MFCD00005156
UN1993
1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64
UKTHULMXFLCNAV-UHFFFAOYSA-N
2-hex-5-enyloxirane
29678
97%

Chemical Identifiers

19600-63-6
126.199
UKTHULMXFLCNAV-UHFFFAOYSA-N
29678
C=CCCCCC1CO1
C8H14O
MFCD00005156
1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64
2-hex-5-enyloxirane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 243-178-7

RTECSNumber : RR0502000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only