Learn More
1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals
$162.60 - $615.36
Chemical Identifiers
CAS | 19600-63-6 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00005156 |
InChI Key | UKTHULMXFLCNAV-UHFFFAOYSA-N |
Synonym | 1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64 |
PubChem CID | 29678 |
IUPAC Name | 2-hex-5-enyloxirane |
SMILES | C=CCCCCC1CO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0703306
|
Thermo Scientific Chemicals
L0703306 |
5 g |
Each for $162.60
|
|
|||||
AAL0703314
|
Thermo Scientific Chemicals
L0703314 |
25 g |
Each for $615.36
|
|
|||||
Description
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
19600-63-6 | |
126.199 | |
UKTHULMXFLCNAV-UHFFFAOYSA-N | |
29678 | |
C=CCCCCC1CO1 |
C8H14O | |
MFCD00005156 | |
1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64 | |
2-hex-5-enyloxirane |
Specifications
19600-63-6 | |
167°C | |
C8H14O | |
MFCD00005156 | |
UN1993 | |
1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64 | |
UKTHULMXFLCNAV-UHFFFAOYSA-N | |
2-hex-5-enyloxirane | |
29678 | |
97% |
0.85 | |
38°C (100°F) | |
1.435 | |
5 g | |
1305290 | |
Not miscible in water. | |
C=CCCCCC1CO1 | |
126.199 | |
126.2 | |
1,2-Epoxy-7-octene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 243-178-7
RTECSNumber : RR0502000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only