missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-O-Isopropylidene-alpha-D-glucofuranose, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1068806

 View more versions of this product

Catalog No. AAL1068806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-O-Isopropylidene-alpha-D-glucofuranose
18549-40-1
C9H16O6
82670
BGGCXQKYCBBHAH-OZRXBMAMSA-N
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
87704
159°C to 160°C
5 g
MFCD00063244
1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose
CC1(OC2C(C(OC2O1)C(CO)O)O)C
220.221
220.22

Chemical Identifiers

18549-40-1
220.221
BGGCXQKYCBBHAH-OZRXBMAMSA-N
87704
CC1(OC2C(C(OC2O1)C(CO)O)O)C
C9H16O6
MFCD00063244
1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 242-420-9

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only