missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-O-Isopropylidene-alpha-D-glucofuranose, Thermo Scientific Chemicals
$53.33 - $189.92
Chemical Identifiers
CAS | 18549-40-1 |
---|---|
Molecular Formula | C9H16O6 |
Molecular Weight (g/mol) | 220.221 |
MDL Number | MFCD00063244 |
InChI Key | BGGCXQKYCBBHAH-OZRXBMAMSA-N |
Synonym | 1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose |
PubChem CID | 87704 |
IUPAC Name | (1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol |
SMILES | CC1(OC2C(C(OC2O1)C(CO)O)O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1068806
|
Thermo Scientific Chemicals
L1068806 |
5 g |
Each for $53.33
|
|
|||||
AAL1068814
|
Thermo Scientific Chemicals
L1068814 |
25 g |
Each for $189.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18549-40-1 | |
220.221 | |
BGGCXQKYCBBHAH-OZRXBMAMSA-N | |
87704 | |
CC1(OC2C(C(OC2O1)C(CO)O)O)C |
C9H16O6 | |
MFCD00063244 | |
1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose | |
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol |
Specifications
159°C to 160°C | |
5 g | |
MFCD00063244 | |
1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose | |
CC1(OC2C(C(OC2O1)C(CO)O)O)C | |
220.221 | |
220.22 |
18549-40-1 | |
C9H16O6 | |
82670 | |
BGGCXQKYCBBHAH-OZRXBMAMSA-N | |
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol | |
87704 | |
1,2-O-Isopropylidene-alpha-D-glucofuranose |
Safety and Handling
EINECSNumber : 242-420-9
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only