missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,5-Trimethoxy-2-nitrobenzene, 98%, Thermo Scientific Chemicals

$214.97 - $2587.92

Chemical Identifiers

CAS 14227-18-0
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00016992
InChI Key VWYAWLZEMLQGJH-UHFFFAOYSA-N
PubChem CID 518903
IUPAC Name 1,3,5-trimethoxy-2-nitrobenzene
SMILES COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1047306
View Documents
Thermo Scientific Chemicals
A1047306
5 g
Each for $214.97
 
AAA1047314
View Documents
Thermo Scientific Chemicals
A1047314
25 g
Each for $814.04
 
AAA1047322
View Documents
Thermo Scientific Chemicals
A1047322
100 g
Each for $2,587.92
 
Description

Description

2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

Solubility
Soluble in water.

Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
Specifications

Chemical Identifiers

14227-18-0
213.189
VWYAWLZEMLQGJH-UHFFFAOYSA-N
1,3,5-trimethoxy-2-nitrobenzene
C9H11NO5
MFCD00016992
518903
COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC

Specifications

14227-18-0
C9H11NO5
5 g
2,4,6-trimethoxynitrobenzene, 1-nitro-2,4,6-trimethoxybenzene, benzene,1,3,5-trimethoxy-2-nitro, benzene, 1,3,5-trimethoxy-2-nitro, acmc-20ap5l, maybridge1_001624, 1,3,5-trimethoxy-2-nitro-benzene, #
VWYAWLZEMLQGJH-UHFFFAOYSA-N
1,3,5-trimethoxy-2-nitrobenzene
518903
98%
150°C to 153°C
MFCD00016992
2277262
Soluble in water.
COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC
213.189
213.19
1,3,5-Trimethoxy-2-nitrobenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only