missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Triphenylformazan 92.0+%, TCI America™
$55.92 - $260.97
Chemical Identifiers
| CAS | 531-52-2 |
|---|---|
| Molecular Formula | C19H16N4 |
| Molecular Weight (g/mol) | 300.365 |
| MDL Number | MFCD00020647 |
| InChI Key | BEIHVSJTPTXQGB-QUHCWSQZSA-N |
| Synonym | Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan |
| PubChem CID | 5464164 |
| IUPAC Name | N'-anilino-N-phenyliminobenzenecarboximidamide |
| SMILES | C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 |
Chemical Identifiers
| 531-52-2 | |
| 300.365 | |
| BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
| 5464164 | |
| C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 |
| C19H16N4 | |
| MFCD00020647 | |
| Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
| N'-anilino-N-phenyliminobenzenecarboximidamide |
Specifications
| 531-52-2 | |
| MFCD00020647 | |
| Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
| C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 | |
| 300.365 | |
| 300.37 | |
| Crystalline Powder |
| C19H16N4 | |
| 1 g | |
| BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
| N'-anilino-N-phenyliminobenzenecarboximidamide | |
| 5464164 | |
| ≥92.0% (HPLC) | |
| 1,3,5-Triphenylformazan |
Safety and Handling
TSCA : Yes