missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3,5-Triphenylformazan 92.0+%, TCI America™
Supplier: TCI America T01751G
Specifications
1,3,5-Triphenylformazan | |
C19H16N4 | |
1 g | |
BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
N'-anilino-N-phenyliminobenzenecarboximidamide | |
5464164 | |
≥92.0% (HPLC) |
531-52-2 | |
MFCD00020647 | |
Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 | |
300.365 | |
300.37 | |
Crystalline Powder |
Chemical Identifiers
531-52-2 | |
300.365 | |
BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
5464164 | |
C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 |
C19H16N4 | |
MFCD00020647 | |
Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
N'-anilino-N-phenyliminobenzenecarboximidamide |
Safety and Handling
TSCA : Yes