missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Diisopropenylbenzene (stabilized with TBC) 97.0+%, TCI America™
$48.44 - $173.02
Chemical Identifiers
CAS | 3748-13-8 |
---|---|
Molecular Formula | C12H14 |
Molecular Weight (g/mol) | 158.244 |
MDL Number | MFCD00008584 |
InChI Key | IBVPVTPPYGGAEL-UHFFFAOYSA-N |
PubChem CID | 77353 |
IUPAC Name | 1,3-bis(prop-1-en-2-yl)benzene |
SMILES | CC(=C)C1=CC(=CC=C1)C(=C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D195725ML
|
TCI America
D195725ML |
25 mL |
Each for $48.44
|
|
|||||
D1957500ML
|
TCI America
D1957500ML |
500 mL |
Each for $173.02
|
|
|||||
Chemical Identifiers
3748-13-8 | |
158.244 | |
IBVPVTPPYGGAEL-UHFFFAOYSA-N | |
1,3-bis(prop-1-en-2-yl)benzene |
C12H14 | |
MFCD00008584 | |
77353 | |
CC(=C)C1=CC(=CC=C1)C(=C)C |
Specifications
3748-13-8 | |
Yellow | |
MFCD00008584 | |
CC(=C)C1=CC(=CC=C1)C(=C)C | |
158.244 | |
158.24 | |
-40°C | |
Liquid |
25 mL | |
C12H14 | |
IBVPVTPPYGGAEL-UHFFFAOYSA-N | |
1,3-bis(prop-1-en-2-yl)benzene | |
77353 | |
≥97.0% (GC) | |
231°C | |
1,3-Diisopropenylbenzene (stabilized with TBC) |
Safety and Handling
RTECSNumber : CY8535000
TSCA : Yes
Recommended Storage : Refrigerator